Ac-LEHD-AFC
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Caspase-9 substrate
Fluorogenic substrate for caspase-9.
Specifications
- Protein/peptide name:Ac-LEHD-AFC
- Purity:>96% (HPLC)
- Formulation:Lyophilized
Specifications
Frequently Bought Together