Order Entry
Poland
ContactUsLinkComponent
Angiotensin IV
Angiotensin IV
numer katalogowy ENZOALX151042M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Angiotensin IV
numer katalogowy ENZOALX151042M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    02
  • Protein/peptide name:
    Angiotensin IV
  • Purity:
    >99%

Specifications

About this item