Order Entry
Poland
ContactUsLinkComponent
BAFF soluble (mouse), detection set
BAFF soluble (mouse), detection set
Catalog # ENZOAPO54N013KI01
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
BAFF soluble (mouse), detection set
Catalog # ENZOAPO54N013KI01
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 SET
  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Conjugate ELISA:
    HRP-Streptavidin
  • Format:
    Uncoated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    BAFF
  • Description:
    BAFF soluble (mouse), detection set
  • Seq:
    02
  • Storage temperature:
    −20 °C
  • Sensitivity:
    0,3 ng/ml (range 0 - 20 ng/ml)
  • Tests per kit:
    4×96 wells

Specifications

About this item

For the quantitative determination of soluble mouse BAFF from biological fluids (serum and cell culture supernatant).

Delivery: Kit contains 1 vial standard (lyophilized) (STD), 1 vial coating antibody (COAT) and 1 vial detection antibody (DET).