Order Entry
Poland
ContactUsLinkComponent
 
DHEA ELISA kit
  ENZOADI900093
New Product
 :  
DHEA ELISA kit
  ENZOADI900093
New Product
 :  

 

  • Pk:
    1 KIT
  • Assay Type:
    Competitive
  • Host:
  • Primary antibody reactivity:
  • Description:
    DHEA ELISA kit
  • Seq:
    02
  • Environmentally Preferable:
  • Sensitivity:
    2,9 pg/ml (range 12,21 - 50,000 pg/ml)

 

 

Highly sensitive ELISA, measuring DHEA in culture supernatants, plasma, serum, saliva, and urine from any species.

For the quantitative determination of DHEA in culture supernatants, plasma, serum, saliva, and urine from any species.

The DHEA EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 5 hours. Absorbance is read at 405 nm. This kit has low reactivity to structurally related molecules and offers sensitive measurement of DHEA in a variety of sample types.

 : Contains GxR IgG Microtiter plate, Assay buffer, Conjugate, Antibody, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-3263 (5x96 wells).