Order Entry
Poland
ContactUsLinkComponent
 
17β-Estradiol ELISA kit
  ENZOADI901008
 :  
17β-Estradiol ELISA kit
  ENZOADI901008
 :  

 

  • Pk:
    1 KIT
  • Assay Type:
    Competitive
  • Target protein:
    17β-Estradiol
  • Description:
    17β-Estradiol ELISA kit
  • Seq:
    02
  • Sensitivity:
    28,5 pg/ml (range 29,3 - 30000 pg/ml)

 

 

Highly sensitive ELISA, measuring as little as 28,5 pg/ml of 17β-Estradiol in < 3 hours.

For the quantitative determination of 17β-Estradiol in culture supernatants and saliva from any species. For serum and plasma use 89141-346.

The 17β-Estradiol EIA kit is a colorimetric competitive enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 405 nm. This kit offers specific and save non-radioactive quantification of estradiol with ready-to-use liquid color-coded reagents to reduce error.

 : Contains GxR Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Steroid displacement reagent. Alternative size available: 89141-060 (1×96 wells).