Anti-14-3-3 zeta Rabbit Polyclonal Antibody
Catalog # ENZOBMLSA4820025
Supplier: ENZO LIFE SCIENCES
Specifications
- Antibody type:Primary
- Antigen symbol:14-3-3 zeta
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Seq:02
- Form:Liquid. Neat serum containing sodium azide.
- Antigen synonyms:FAS|143Z|KCIP-1|YWHAZ|Factor activating exoenzyme S|1433Z|Protein kinase C inhibitor protein-1|Mitochondrial import stimulation factor S1 subunit|14-3-3 protein zeta/delta
- Pk:25 µl
Specifications
About this item
Host: Rabbit
- For Immunohistochemistry, Western Blot, Affinity Chromatography
Recognizes human, mouse, rat, cow, sheep and chicken 14-3-3zeta.
Type: Primary
Antigen: 14-3-3 zeta
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human