Anti-VASP Mouse Monoclonal Antibody (Biotin) [clone: ]
Catalog # ENZOALX804240BC100
Supplier: Enzo Life Sciences
Specifications
- Antibody type:Primary
- Antigen name:Vasodilator-stimulated phosphoprotein
- Antigen symbol:VASP
- Clonality:Monoclonal
- Conjugation:Biotin
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Seq:02
- Form:Liquid. 0,5 mg/ml in 2X PBS containing 0,09% sodium azide, PEG and sucrose.
- Concentration:0,5 mg/ml
- Purification:Thiophilic adsorption and size exclusion chromatography purified
- Pk:100 µG
Specifications
About this item
Host: Mouse, Isotype: IgG1
- For ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot
Recognizes human, mouse, rat and rabbit Ser239-phosphorylated VASP. Does not cross-react with the non-phosphorylated form of VASP.
Type: Primary
Antigen: VASP
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human