Anti-MAPK Mouse Monoclonal Antibody [clone: 12D4]
Catalog # ENZOALX804061C100
Supplier: Enzo Life Sciences
Specifications
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase
- Antigen symbol:MAPK
- Clonality:Monoclonal
- Clone:12D4
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Dog,Rat,Mouse
- Western blot:Yes
- Seq:02
- Form:Lyophilised from 1 ml of 2X PBS containing 0,09% sodium azide, PEG, and sucrose.
- Antigen synonyms:Extracellular signal regulated kinase 1|Insulin stimulated MAP2 kinase|ERK1|MAP kinase 1|Microtubule associated protein 2 ki|ERT2|MAPK1
- Purification:Thiophilic adsorption and size exclusion chromatography purified
- Pk:100 µG
Specifications
About this item
Host: Mouse, Isotype: IgG1
- For ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Recognizes the pThr-Glu-pTyr motif of activated human, mouse, rat and dog ERK1 and ERK2. Requires phosphorylation both at the threonine and the tyrosine site and does not cross-react with the non-phosphorylated form of the protein. Does not cross-react with activated JNK or p38.
Reconstitute with 1ml water (15 minutes at room temperature).
Type: Primary
Antigen: MAPK
Clonality: Monoclonal
Clone: 12D4
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Mouse, Rat