Order Entry
Poland
ContactUsLinkComponent
5-Norbornene-2-methanol (endo + exo mixture) ≥98%, Sigma-Aldrich®
 
5-Norbornene-2-methanol (endo + exo mixture) ≥98%, Sigma-Aldrich®
  SIAL248533-5G
 :  Merck
 :  
 
5-Norbornene-2-methanol (endo + exo mixture) ≥98%, Sigma-Aldrich®
  SIAL248533-5G
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Assay:
    98%
  • Refractive index n20/D:
    1,500 (l)
  • Density (25 °C):
    1,027 g/ml

 

 

 :
 :124.183 g/mol
 :97 °C (20 mmHg) (l)
 :1.027 g/ml (25 °C)
 :M-Clarity™ quality level = MQ100
 :87 °C
 :GHS07
 :95-12-5
 :C₈H₁₂O
 :MFCD00167571