Order Entry
Poland
ContactUsLinkComponent
AmberChrom™ chloride form, 1X8 50 -100 Mesh, Supelco®
AmberChrom™ chloride form, 1X8 50 -100 Mesh, Supelco®
Supplier:  Merck
Certificates
undefined
AmberChrom™ chloride form, 1X8 50 -100 Mesh, Supelco®
Supplier:  Merck

Specification Test Results

  • Form:
    Liquid
  • Colour:
    Almost colourless to yellow and almost colourless to green-yellow
  • Purity (GC):
    ≥98,5%
  • Infrared spectrum:
    Conforms to structure

Specification Test Results

Frequently Bought Together