Order Entry
Poland
ContactUsLinkComponent
Anti-Tuberin Polyclonal Antibody
Anti-Tuberin Polyclonal Antibody
Catalog # BSBTPB9121
Supplier:  Boster Bio
undefined
Anti-Tuberin Polyclonal Antibody
Catalog # BSBTPB9121
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Tuberin
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Polyclonal antibody for P TUBERIN/TSC2 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. P TUBERIN/TSC2 information: Molecular Weight: 200608 MW; Subcellular Localization: Cytoplasm. Membrane; Peripheral membrane protein. At steady state found in association with membranes; Tissue Specificity: Liver, brain, heart, lymphocytes, fibroblasts, biliary epithelium, pancreas, skeletal muscle, kidney, lung and placenta.

Type: Primary
Antigen: Tuberin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat