Order Entry
Poland
ContactUsLinkComponent
Anti-MSH2 Polyclonal Antibody
Anti-MSH2 Polyclonal Antibody
Catalog # BSBTPB9284
Supplier:  Boster Bio
undefined
Anti-MSH2 Polyclonal Antibody
Catalog # BSBTPB9284
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    MSH2
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Polyclonal antibody for MSH2 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. MSH2 information: Molecular Weight: 104743 MW; Subcellular Localization: Nucleus ; Tissue Specificity: Ubiquitously expressed.

Type: Primary
Antigen: MSH2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat