Anti-Hamartin Polyclonal Antibody
Catalog # BSBTPB9120
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Hamartin
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for HAMARTIN/TSC1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. HAMARTIN/TSC1 information: Molecular Weight: 129767 MW; Subcellular Localization: Cytoplasm . Membrane ; Peripheral membrane protein . At steady state found in association with membranes; Tissue Specificity: Highly expressed in skeletal muscle, followed by heart, brain, placenta, pancreas, lung, liver and kidney. Also expressed in embryonic kidney cells.
Type: Primary
Antigen: Hamartin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Rat