Anti-Egr1 Polyclonal Antibody
Catalog # BSBTA00687-1
Supplier: Boster Bio
Specifications
- Typ przeciwciała:Primary
- Symbol antygenu:Egr1
- Klonalność:Poliklonalne
- Gospodarz:Rabbit
- Reaktywność:Human,Rat,Mouse
- Rozmiar:100 µg
- Ilość:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Egr1 detection. Tested with WB, Direct ELISA in Human;Mouse;Rat.
Type: Primary
Antigen: Egr1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat