Anti-Cyclin D3 Polyclonal Antibody
Catalog # BSBTPB9103
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:Cyclin D3
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for Cyclin D3/CCND3 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. Cyclin D3/CCND3 information: Molecular Weight: 32520 MW; Subcellular Localization: Nucleus . Cytoplasm . Membrane . Cyclin D-CDK4 complexes accumulate at the nuclear membrane and are then translocated to the nucleus through interaction with KIP/CIP family members.
Type: Primary
Antigen: Cyclin D3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Rat