Anti-Claudin 2 Polyclonal Antibody
Catalog # BSBTRP1042
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:Claudin 2
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for CLAUDIN 2/CLDN2 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. CLAUDIN 2/CLDN2 information: Molecular Weight: 24549 MW; Subcellular Localization: Cell junction, tight junction . Cell membrane ; Multi-pass membrane protein .
Type: Primary
Antigen: Claudin 2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Rat