Anti-CD43 Polyclonal Antibody
Catalog # BSBTPB9095
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:CD43
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for CD43/SPN detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. CD43/SPN information: Molecular Weight: 40322 MW; Subcellular Localization: Membrane; Single-pass type I membrane protein; Tissue Specificity: Cell surface of thymocytes, T-lymphocytes, neutrophils, plasma cells and myelomas.
Type: Primary
Antigen: CD43
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat