Anti-Caspase 8 Polyclonal Antibody
BSBTA00042-1
- Antibody type:Primary
- Antigen name:Caspase 8
- Antigen symbol:CASP8
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for Caspase 8 detection. Tested with WB, Direct ELISA in Human;Mouse;Rat.
Type: Primary
Antigen: Caspase 8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat