4-Azabenzimidazole formate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
:
Formula:C₆H₅N₃·CH₂O₂
MDL Number:MFCD30145860
:165.15 g/mol
Melting Pt:77…78 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:Yellow to brown Powder
- Infrared spectrum:Conforms
- NMR (1H-NMR):Consistent with structure
- Elemental Analysis:≥48.87 to ≤52.95 % (Carbon)
Specification Test Results
Frequently Bought Together