Order Entry
Poland
ContactUsLinkComponent
 
Irritant
4-Chloro-2-fluoro-1-nitrobenzene 98%
Catalog # ACRO455930250
CAS Number:  
undefined
Irritant
4-Chloro-2-fluoro-1-nitrobenzene 98%
Catalog # ACRO455930250
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Off-white to yellow Low melting solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    46 to 52 °C
  • GC:
    ≥97.5 %

Specification Test Results

About this item

 :
 :175.55 g/mol
Melting Pt:48…50 °C
Flash Pt:>110 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:700-37-8
Formula:C₆H₃ClFNO₂
MDL Number:MFCD00042211