Order Entry
Poland
ContactUsLinkComponent
Thapsigargin ≥97% (by TLC), colorless solid, Millipore®
Thapsigargin ≥97% (by TLC), colorless solid, Millipore®
Catalog # 586005-1
CAS Number:  
Thapsigargin ≥97% (by TLC), colorless solid, Millipore®
Catalog # 586005-1
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :650.762 g/mol
Storage Temperature:Freezer
CAS Number:67526-95-8
Formula:C₃₄H₅₀O₁₂
MDL Number:MFCD00083511