Supelco® Drug and metabolite standards, theobromine, LoGiCal®
Catalog # PROOLGCFOR0577.00
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:10 mg
- Standards:Reference standard
- CAS No.:83-67-0
- Description:Theobromine
- Formula:C₇H₈N₄O₂
- Description:Theobromine