Supelco® Drug and metabolite standards, Sildenafil N-Oxide, LoGiCal®
Catalog # PROOLGCFOR0254.04
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:10 mg
- Standards:Reference standard
- CAS No.:1094598-75-0
- Description:Sildenafil N-oxide
- Formula:C₂₂H₃₀N₆O₅S
- Description:Sildenafil N-oxide