Order Entry
Poland
Orders LinkContactUsLinkComponent
Ethinyl Estradiol, Reference Standards

Ethinyl Estradiol, Reference Standards

Supplier:   USP
Ethinyl Estradiol, Reference Standards
Supplier:  USP
Certificates

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1260001.
  • 1260012.
  • 1260023.
  • Description:
  • Ethinyl Estradiol
  • Ethinyl Estradiol Related Compound A
  • Ethinyl Estradiol Related Compound B
  • Synonym(s):
  • 19-Nor-17α-pregna-1,3,5(10)-trien-20-yne-3,17-diol-6-one
  • 19-Nor-17α-pregna-1,3,5(10),9(11)-tetraen-20-yne-3,17-diol monohydrate
  • CAS No.:
  • 57-63-6
  • 38002-18-5
  • 1231-96-5
  • Formula:
  • C₂₀H₂₄O₂
  • C₂₀H₂₂O₃
  • C₂₀H₂₂O₂
  • UN No.:
  • 3077

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1260001.
    Ethinyl Estradiol
    Ethinyl Estradiol
    USP reference standard
    Vial
    150 mg
    Log in for availability
    Log in for availability
    1 * 150 mg
    1 * 150 mg - 2 010 zł
    2 010 zł
    1 * 150 mg
       2 010 zł

    Specifications:
    More
    Specifications
    Cat. No.
    1260001.
    Description
    Ethinyl Estradiol
    Synonym(s)
    CAS No.
    57-63-6
    Formula
    C₂₀H₂₄O₂
    UN No.
    3077
  • 1260012.
    Ethinyl Estradiol Related Compound A
    Ethinyl Estradiol Related Compound A
    USP reference standard
    Vial
    20 mg
    Log in for availability
    Log in for availability
    1 * 20 mg
    1 * 20 mg - 5 780 zł
    5 780 zł
    1 * 20 mg
       5 780 zł

    Specifications:
    More
    Specifications
    Cat. No.
    1260012.
    Description
    Ethinyl Estradiol Related Compound A
    Synonym(s)
    19-Nor-17α-pregna-1,3,5(10)-trien-20-yne-3,17-diol-6-one
    CAS No.
    38002-18-5
    Formula
    C₂₀H₂₂O₃
    UN No.
  • 1260023.
    Ethinyl Estradiol Related Compound B
    Ethinyl Estradiol Related Compound B
    USP reference standard
    Vial
    20 mg
    Log in for availability
    Log in for availability
    1 * 20 mg
    1 * 20 mg - 6 120 zł
    6 120 zł
    1 * 20 mg
       6 120 zł

    Specifications:
    More
    Specifications
    Cat. No.
    1260023.
    Description
    Ethinyl Estradiol Related Compound B
    Synonym(s)
    19-Nor-17α-pregna-1,3,5(10),9(11)-tetraen-20-yne-3,17-diol monohydrate
    CAS No.
    1231-96-5
    Formula
    C₂₀H₂₂O₂
    UN No.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.