Organic reference standard, Meclozine dihydrochloride, Ph.Eur., RS
Catalog # EDQMM0220000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:150 mg
- Standards:Reference standard
- CAS No.:1104-22-9
- Description:Meclozine dihydrochloride
- Environmentally Preferable:
- Formula:C₂₅H₂₉Cl₃N₂
- Storage temperature:Ambient
- Description:Meclozine dihydrochloride
Specifications
About this item
- In Monograph 0622
- Solubility in water: 1 to 10 g/l