Organic reference standard, Dexpanthenol, Ph.Eur., RS
Catalog # EDQMD0730000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:5 mg
- Standards:Reference standard
- CAS No.:81-13-0
- Description:Dexpanthenol
- Formula:C₉H₁₉NO₄
- Storage temperature:Ambient
- Description:Dexpanthenol
Specifications
About this item
- In Monograph 0761
- Soluble in/miscible with alcohol, propylene, glycol, and water
- Pharmaceutical industry applications