Order Entry
Poland
Orders LinkContactUsLinkComponent
Mupirocin, Reference Standards

Mupirocin, Reference Standards

Supplier:   USP
Mupirocin, Reference Standards
Supplier:  USP
Certificates

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1448901.
  • 1448923.
  • 1448912.
  • Description:
  • Mupirocin
  • Mupirocin Lithium
  • Mupirocin Calcium
  • CAS No.:
  • 12650-69-0
  • 73346-79-9
  • 115074-43-6
  • Formula:
  • C₂₆H₄₄O₉
  • C₂₆H₄₃LiO₉
  • C₅₂H₉₀CaO₂₀

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1448901.
    Mupirocin
    Mupirocin
    USP reference standard
    Vial
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 2 010 zł
    2 010 zł
    1 * 50 mg
       2 010 zł

    Specifications:
    More
    Specifications
    Cat. No.
    1448901.
    Description
    Mupirocin
    CAS No.
    12650-69-0
    Formula
    C₂₆H₄₄O₉
  • 1448923.
    Mupirocin Lithium
    Mupirocin Lithium
    USP reference standard
    Vial
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 2 430 zł
    2 430 zł
    1 * 100 mg
       2 430 zł

    Specifications:
    More
    Specifications
    Cat. No.
    1448923.
    Description
    Mupirocin Lithium
    CAS No.
    73346-79-9
    Formula
    C₂₆H₄₃LiO₉
  • 1448912.
    Mupirocin Calcium
    Mupirocin Calcium
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 2 570 zł
    2 570 zł
    1 * 200 mg
       2 570 zł

    Specifications:
    More
    Specifications
    Cat. No.
    1448912.
    Description
    Mupirocin Calcium
    CAS No.
    115074-43-6
    Formula
    C₅₂H₉₀CaO₂₀
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.