Order Entry
Poland
Orders LinkContactUsLinkComponent
Drospirenone, Reference Standards

Drospirenone, Reference Standards

Supplier:   USP
Drospirenone, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1229409.
  • 1229410.
  • Description:
  • Drospirenone
  • Drospirenone Related Compound A
  • Synonym(s):
  • 17-Hydroxy-6β,7β:15β,16β-dimethylene-3-oxo-17β-pregn-4-ene-21-carboxylic acid, gamma-lactone
  • CAS No.:
  • 67392-87-4
  • 90457-65-1
  • Formula:
  • C₂₄H₃₀O₃
  • C₂₄H₃₀O₃

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1229409.
    Drospirenone
    Drospirenone
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 2 010 zł
    2 010 zł
    1 * 200 mg
       2 010 zł

    Specifications:
    More
    Specifications
    Cat. No.
    1229409.
    Description
    Drospirenone
    Synonym(s)
    CAS No.
    67392-87-4
    Formula
    C₂₄H₃₀O₃
  • 1229410.
    Drospirenone Related Compound A
    Drospirenone Related Compound A
    USP reference standard
    Vial
    15 mg
    Log in for availability
    Log in for availability
    1 * 15 mg
    1 * 15 mg - 5 940 zł
    5 940 zł
    1 * 15 mg
       5 940 zł

    Specifications:
    More
    Specifications
    Cat. No.
    1229410.
    Description
    Drospirenone Related Compound A
    Synonym(s)
    17-Hydroxy-6β,7β:15β,16β-dimethylene-3-oxo-17β-pregn-4-ene-21-carboxylic acid, gamma-lactone
    CAS No.
    90457-65-1
    Formula
    C₂₄H₃₀O₃
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.