Order Entry
Poland
Orders LinkContactUsLinkComponent
Docetaxel, Reference Standards

Docetaxel, Reference Standards

Supplier:   USP
Docetaxel, Reference Standards
Supplier:  USP
Certificates

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1224551.
  • 1224562.
  • 1224573.
  • Description:
  • Docetaxel
  • Docetaxel Identication
  • Docetaxel System Suitability Mixture
  • Synonym(s):
  • (2R,3S)-N-Carboxy-3-phenylisoserine, N-tert-butyl ester, 13-ester with 5β,20-epoxy-1,2α,4,7β,10β,13α-hexahydroxytax-11-en-9-one 4-acetate 2-benzoate trihydrate
  • CAS No.:
  • 148408-66-6
  • Formula:
  • C₄₃H₅₉NO₁₇

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1224551.
    Docetaxel
    Docetaxel
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 14 230 zł
    14 230 zł
    1 * 200 mg
       14 230 zł

    Specifications:
    More
    Specifications
    Cat. No.
    1224551.
    Description
    Docetaxel
    Synonym(s)
    (2R,3S)-N-Carboxy-3-phenylisoserine, N-tert-butyl ester, 13-ester with 5β,20-epoxy-1,2α,4,7β,10β,13α-hexahydroxytax-11-en-9-one 4-acetate 2-benzoate trihydrate
    CAS No.
    148408-66-6
    Formula
    C₄₃H₅₉NO₁₇
  • 1224562.
    Docetaxel Identication
    Docetaxel Identication
    USP reference standard
    Vial
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 6 550 zł
    6 550 zł
    1 * 50 mg
       6 550 zł

    Specifications:
    More
    Specifications
    Cat. No.
    1224562.
    Description
    Docetaxel Identication
    Synonym(s)
    CAS No.
    Formula
  • 1224573.
    Docetaxel System Suitability Mixture
    Docetaxel System Suitability Mixture
    USP reference standard
    Vial
    15 mg
    Log in for availability
    Log in for availability
    1 * 15 mg
    1 * 15 mg - 6 640 zł
    6 640 zł
    1 * 15 mg
       6 640 zł

    Specifications:
    More
    Specifications
    Cat. No.
    1224573.
    Description
    Docetaxel System Suitability Mixture
    Synonym(s)
    CAS No.
    Formula