Order Entry
Poland
ContactUsLinkComponent
Goserelin Related Compounds, Reference Standard
Goserelin Related Compounds, Reference Standard
Supplier:  USP
Certificates
Goserelin Related Compounds, Reference Standard
Supplier:  USP

Specifications

  • CAS No.:
    145781-92-6
  • Formula:
    C₂₄H₂₇NO₃
  • Description:
    Goserelin Acetate
  • Synonym(s):
    4-D-Serine Goserelin, 1-(5-Oxo-L-prolyl-L-histidyl-L-tryptophyl-D-seryl-L-tyrosyl-O-tert-butyl-D-seryl-L-leucyl-L-arginyl-L-prolyl) semicarbazide

Specifications

Frequently Bought Together