Order Entry
Poland
Orders LinkContactUsLinkComponent
Lomustine, Reference Standards

Lomustine, Reference Standards

Supplier:   USP
Lomustine, Reference Standards
Supplier:  USP
Certificates

4 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1369419.
  • 1369442.
  • 1369453.
  • 1369431.
  • Description:
  • Lomustine
  • Lomustine Related Compound C
  • Lomustine Related Compound D
  • Lomustine Related Compound B
  • Synonym(s):
  • 1,3-Dicyclohexylurea
  • 3-(2-Chloroethyl)-1-cyclohexyl-1-nitrosourea
  • 1-(2-Chloroethyl)-3-cyclohexylurea
  • CAS No.:
  • 13010-47-4
  • 2387-23-7
  • 54749-91-6
  • 13010-47-4
  • Formula:
  • C₉H₁₆ClN₃O₂
  • C₁₃H₂₄N₂O
  • C₉H₁₆ClN₃O₂
  • UN No.:
  • 2811

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1369419.
    Lomustine
    Lomustine
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 2 130 zł
    2 130 zł
    1 * 200 mg
       2 130 zł

    Specifications:
    More
    Specifications
    Cat. No.
    1369419.
    Description
    Lomustine
    Synonym(s)
    CAS No.
    13010-47-4
    Formula
    C₉H₁₆ClN₃O₂
    UN No.
    2811
  • 1369442.
    Lomustine Related Compound C
    Lomustine Related Compound C
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 6 480 zł
    6 480 zł
    1 * 25 mg
       6 480 zł

    Specifications:
    More
    Specifications
    Cat. No.
    1369442.
    Description
    Lomustine Related Compound C
    Synonym(s)
    1,3-Dicyclohexylurea
    CAS No.
    2387-23-7
    Formula
    C₁₃H₂₄N₂O
    UN No.
  • 1369453.
    Lomustine Related Compound D
    Lomustine Related Compound D
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 6 400 zł
    6 400 zł
    1 * 25 mg
       6 400 zł

    Specifications:
    More
    Specifications
    Cat. No.
    1369453.
    Description
    Lomustine Related Compound D
    Synonym(s)
    3-(2-Chloroethyl)-1-cyclohexyl-1-nitrosourea
    CAS No.
    54749-91-6
    Formula
    C₉H₁₆ClN₃O₂
    UN No.
  • 1369431.
    Lomustine Related Compound B
    Lomustine Related Compound B
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 6 540 zł
    6 540 zł
    1 * 25 mg
       6 540 zł

    Specifications:
    More
    Specifications
    Cat. No.
    1369431.
    Description
    Lomustine Related Compound B
    Synonym(s)
    1-(2-Chloroethyl)-3-cyclohexylurea
    CAS No.
    13010-47-4
    Formula
    UN No.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.