Order Entry
Poland
ContactUsLinkComponent
 
Human recombinant IL-2 R beta (from cells)
  PRSI92-480
 :  
Human recombinant IL-2 R beta (from cells)
  PRSI92-480
 :  

 

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-Fc tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    High affinity IL-2 receptor subunit beta|IL-2RB|CD122|IL-2R subunit beta|Interleukin-2 receptor subunit beta|IL2RB|IL-2 receptor subunit beta
  • Protein/peptide name:
    IL-2 R beta
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    51.7 kD
  • Sequence:
    Ala28-Leu239
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.

 

 

Human IL-2RB, also known asinterleukin-2 receptor subunit beta,is the receptor for interleukin-2. IL2 receptor complex is involved in receptor mediated endocytosis and transduces the mitogenic signals of IL2. IL2 receptor complex has three forms with respect to ability to bind IL2. IL-2RB is belonged to a type I membrane protein,and has a 26 residue signal peptide, a 214 residue extracellular region, a 25 residue transmembrane region and a 286 residue cytoplasmic domain. IL-2RB is the subunit critical for receptor-mediated signalling via physically or functionally coupling to other signalling molecules, such as the Jak-STAT and Src-family protein tyrosine kinase although it lacks apparent catalytic motifs.

Fusion-Tag: C-Fc tag

This recombinant protein can be used for biological assays. For research use only.