Human recombinant IL-1 Rrp2 (from Cells)
Catalog # PRSI92-460
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-Fc tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:IL-36 receptor|IL1RL2|IL-1RL2|IL1R-rp2|IL-36R|IL-1Rrp2|IL-1 R6|Interleukin-1 receptor-like 2
- Protein/peptide name:IL-1 Rrp2
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:78.8 kD
- Sequence:Met1-Gly457
- Formulation:Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
Specifications
About this item
Interleukin-1 receptor-like 2 is a protein that in humans is encoded by the IL1RL2 gene, belongs to the interleukin-1 receptor family.IL1RL2 is the receptor for interleukin-36 (IL36A, IL36B and IL36G). After binding to interleukin-36 associates with the coreceptor IL1RAP to form the interleukin-36 receptor complex which mediates interleukin-36-dependent activation of NF-kappa-B, MAPK and other pathways.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.