Order Entry
Poland
ContactUsLinkComponent
Human recombinant MMP9 (from cells)
Human recombinant MMP9 (from cells)
Catalog # PRSI92-376
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant MMP9 (from cells)
Catalog # PRSI92-376
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    92 kDa gelatinase|MMP9|Matrix metalloproteinase-9|92 kDa type IV collagenase|Gelatinase B
  • Protein/peptide name:
    MMP9
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    77.4 kD
  • Sequence:
    Ala19-Asp707
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 2mM CaCl2, 150mM NaCl, 0.05% Brij35(w/v), pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Matrix metallopeptidase 9 (MMP-9) is an enzyme encoded by the MMP9 gene. This protein, which is produced by normal alveolar macrophages and granulocytes, can be activated by 4-aminophenylmercuric acetate and phorbol ester and up-regulated by ARHGEF4, SPATA13 and APC via the JNK signalling pathway in colorectal tumour cells. MMP-9 is involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, angiogenesis, bone development, wound healling, cell migration, learning and memory, as well as in pathological processes, such as arthritis, intracerebral hemorrhage, and metastasis.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.