Order Entry
Poland
ContactUsLinkComponent
Human Recombinant 40S Ribosomal S7 (from E. coli)
Human Recombinant 40S Ribosomal S7 (from E. coli)
Catalog # PRSI92-289
Supplier:  ProSci Inc.
CAS Number:  
Human Recombinant 40S Ribosomal S7 (from E. coli)
Catalog # PRSI92-289
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    RPS7|40S ribosomal protein S7
  • Protein/peptide name:
    40S Ribosomal S7
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    23.2 kD
  • Sequence:
    Met1-Leu194
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM Tris,150mM NaCl,pH8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

40S ribosomal protein S7(RPS7) belongs to the S7E family of ribosomal proteins. It is phosphorylated by NEK6 during post-translational modification. RPS7 is located in the cytoplasm, binds IPO9 with high affinity. it also can interacts with NEK6. As is required for rRNA maturation and typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. The unnormal expression of RPS7 may cause Diamond-Blackfan anemia 8.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.