Order Entry
Poland
ContactUsLinkComponent
Human recombinant IL21 receptor (from cells)
Human recombinant IL21 receptor (from cells)
Catalog # PRSI91-859
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant IL21 receptor (from cells)
Catalog # PRSI91-859
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    IL-21R|CD360.|Interleukin-21 receptor|IL-21 receptor
  • Protein/peptide name:
    IL21 Receptor
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    26 kD
  • Sequence:
    Cys20-Pro236
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Interleukin-21 receptor is also known as IL-21 receptor, IL-21R, CD360. In humans, it is encoded by the IL21R gene. It belongs to the type I cytokine receptor family. Type 4 subfamily contains 2 fibronectin type-III domains. Interleukin-21 receptor is selectively expressed in lymphoid tissues and highly expressed in thymus and spleen. IL-21 is produced by CD4+ T cells in response to antigenic stimulation. Its action enhances antigen-specific responses of immune cells. The biological effects of IL-21 include induction of differentiation of T-cells-stimulated B-cells into plasma cells and memory B-cells. It also promotes the anti-tumor activity of CD8+ T-cells and NK cells.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag