Order Entry
Poland
ContactUsLinkComponent
Human recombinant WARS (from E. coli)
Human recombinant WARS (from E. coli)
Catalog # PRSI91-280
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant WARS (from E. coli)
Catalog # PRSI91-280
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Interferon-induced protein 53.|TrpRS|WARS also known as Tryptophanyl-tRNA synthetase
  • Protein/peptide name:
    WARS
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    55.3 kD
  • Sequence:
    Met1-Gln471
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM Tris, 150mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

There exists two types of tryptophanyl tRNA synthetases, the cytoplasmic form called WARS, the mitochondrial form called WARS2. WARS catalyzes the aminoacylation of tRNA (trp) with tryptophan and is induced by interferon. WARS regulates ERK, Akt, eNOS activation pathway, which are related with angiogenesis, cytoskelatal reorganization and shear stess-reponsive gene expression.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: N-6 His tag