Human recombinant Annexin A13 (from E. coli)
Catalog # PRSI91-184
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:Annexin A13|Annexin-13|ANXA13|Intestine-Specific Annexin|ISA|ANX13|Annexin XIII
- Protein/peptide name:Annexin A13
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:35.4 kD
- Sequence:Gly2-His316
- Formulation:Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 100mM NaCl, 10% Glycerol, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Annexin A13 (ANXA13) belongs to the annexin family which plays a role in phospholipase inhibition, cytoskeletal interactions, intracellular signal transduction pathways and regulation of cellular growth. ANXA13 contains four annexin repeats and a pair of annexin repeats may form one binding site for calcium and phospholipid. ANXA13 is highly expressed in intestinal and kidney epithelial cells. The specific function of ANXA13 has not yet been determined; however it is associated with the plasma membrane of undifferentiated, proliferating crypt epithelial cells as well as differentiated villus enterocytes.
- This recombinant protein can be used for biological assays. For research use only.