Human recombinant REG4 (from HEK293 cells)
PRSI96-644
:
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Protein synonyms:REG-IV|REG4|GISP|RELP
- Protein/peptide name:REG4
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:16.7 kDa
- Formulation:Lyophilized, PBS, pH7.4
- Tested applications:Western Blot
Regenerating islet-derived protein 4 (REG4) is also known as Gastrointestinal secretory protein (GISP), REG-like protein (RELP). REG4 contains one C-type lectin domain. REG4 has calcium-independent lectin displaying mannose-binding specificity and able to maintain carbohydrate recognition activity in an acidic environment. REG4 may be involved in inflammatory and metaplastic responses of the gastrointestinal epithelium.