Human recombinant CD55 (from HEK293 cells)
Catalog # PRSI96-117
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Biological activity:Measured by its binding ability in a functional ELISA. When rhCD55 is coated at 1μg/ml (100μl/well), the concentration of rhCD97 that produces 50% of the optimal binding response is found to be approximately 0. 15-2. 0μg/ml.
- Protein synonyms:CR|DAF|CD55|CROM|TC
- Protein/peptide name:CD55
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:35.8 kDa
- Formulation:Lyophilized, PBS, pH7.4
- Tested applications:Western Blot