Human RNF8 peptide (14 aa near C-terminus)
PRSI5763P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:14 aa near C-terminus
- Protein/peptide name:RNF8 Peptide (14 aa near C-terminus)
- Tested applications:Blocking
RNF8 peptide is used for blocking the activity of RNF8 antibody.