Human Dact3 peptide (17 aa near N-terminus)
PRSI4871P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:17 aa near N-terminus
- Protein/peptide name:Dact3 Peptide (17 aa near N-terminus)
- Tested applications:Blocking
Dact3 peptide is used for blocking the activity of Dact3 antibody.