Order Entry
Poland
ContactUsLinkComponent
 
Human OTUD5 peptide (13 aa near C-terminus)
  PRSI4811P
 :  
Human OTUD5 peptide (13 aa near C-terminus)
  PRSI4811P
 :  

 

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    13 aa near C-terminus
  • Protein/peptide name:
    OTUD5 Peptide (13 aa near C-terminus)
  • Tested applications:
    Blocking

 

 

OTUD5 peptide is used for blocking the activity of OTUD5 antibody.