Human ACVR1B peptide (14 aa near C-terminus)
PRSI4793P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:14 aa near C-terminus
- Protein/peptide name:ACVR1B Peptide (14 aa near C-terminus)
- Tested applications:Blocking
ACVR1B peptide is used for blocking the activity of ACVR1B antibody.