Human ORAI2 peptide (15 aa near C-terminus)
Catalog # PRSI4111P
Dostawca: ProSci Inc.
Numer CAS:
Dane techniczne
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:15 aa near C-terminus
- Protein/peptide name:ORAI2 Peptide (15 aa near C-terminus)
- Tested applications:Blocking
Dane techniczne
Informacje o tym produkcie
ORAI2 peptide is used for blocking the activity of ORAI2 antibody.