Human TIM-4 peptide (15 aa near C-terminus)
PRSI3813P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:15 aa near C-terminus
- Protein/peptide name:TIM-4 Peptide (15 aa near C-terminus)
- Tested applications:Blocking
TIM-4 peptide is used for blocking activity of TIM-4 antibody.