Order Entry
Poland
ContactUsLinkComponent
Human IL-21 receptor peptide (15 aa near the extracellualr domain)
Human IL-21 receptor peptide (15 aa near the extracellualr domain)
Catalog # PRSI2469P
Supplier:  ProSci Inc.
CAS Number:  
Human IL-21 receptor peptide (15 aa near the extracellualr domain)
Catalog # PRSI2469P
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    15 aa near the extracellualr domain
  • Protein/peptide name:
    IL-21 Receptor Peptide (15 aa near the extracellualr domain)
  • Tested applications:
    Blocking

Specifications

About this item

IL-21 receptor peptide is used for blocking the activity of IL-21 receptor antibody.