Anti-STAT5A/STAT5B Rabbit Polyclonal Antibody
Catalog # PRSI80-026
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Signal Transducer and Activator of Transcription 5A, Signal Transducer and Activator of Transcription 5B
- Antigen symbol:STAT5A/STAT5B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:6776;6777
- Antigen synonyms:STAT5|STA5B|Signal transducer and activator of transcription 5B|STAT5A|transcription factor STAT5B|MGF
- Modification:phospho-Ser725/730
- Storage buffer:Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
- Molecular weight:90 kDa
- Storage temperature:Store antibody at –20 °C for up to one year.
- Concentration:1 mg/mL
- Shipping temperature:-20°C
- Immunogen:STAT5A/B (phospho Ser725/730) antibody was raised against a peptide sequence around phosphorylation site of serine725/730 (A-P-S (p)-P-A) derived from Human STAT5A/B .
- Purification:Purified by affinity-chromatography using epitope-specific peptide.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Carries out a dual function: signal transduction and activation of transcription. Binds to the GAS element and activates PRL-induced transcription.
Western Blot: 1:500~1:1000
Type: Primary
Antigen: STAT5A/STAT5B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat