Order Entry
Poland
ContactUsLinkComponent
Anti-UBC Rabbit Polyclonal Antibody
Anti-UBC Rabbit Polyclonal Antibody
Catalog # PRSI70-905
Supplier:  ProSci Inc.
undefined
Anti-UBC Rabbit Polyclonal Antibody
Catalog # PRSI70-905
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Ubiquitin C
  • Antigen symbol:
    UBC
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Antigen synonyms:
    HMG20
  • Concentration:
    0.25 mg/mL
  • Immunogen:
    KLH conjugated synthetic peptide selected from the C-Terminal region of human Ubiquitin.
  • Size:
    50 µg
  • Pk:
    200 µl

Specifications

About this item

FABP4 Antibody can be used for ELISA starting at 1:1000, and Immunohistochemistry in paraffin embeded tissues starting at 5 μg/mL,

Protein G Column

PBS, 0.09% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: UBC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human