Order Entry
Poland
ContactUsLinkComponent
Anti-SLC2A2 Mouse Monoclonal Antibody
Anti-SLC2A2 Mouse Monoclonal Antibody
Catalog # PRSI49-922
Supplier:  ProSci Inc.
undefined
Anti-SLC2A2 Mouse Monoclonal Antibody
Catalog # PRSI49-922
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Solute Carrier Family 2 Member A2
  • Antigen symbol:
    SLC2A2
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Form:
    Liquid
  • Antigen synonyms:
    GLUT2
  • Immunogen:
    SLC2A2 monoclonal antibody was raised against glut2 - transfected NS0 cells (Human).
  • Tested applications:
    Fluorescence-Activated Cell Sorting
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Glut2 belongs to the facilitative glucose transporter protein family that comprises 13 members. It is an integral membrane protein with 12 transmembrane domains. Glut2 is expressed predominantly in liver, intestine, kidney, and pancreatic beta-cells. It is a low-affinity glucose transporter that has been suggested to function as a glucose sensor in pancreatic beta-cells and facilitate either glucose uptake or efflux from cells depending on the nutritional state.1 Suitable for use in Flow Cytometry and Immunohistochemistry. Other applications not tested. Recognizes human GLUT 2 (Glucose Transporter, Insulin Regulatable) expression on hGlut2-transfected NS0 cells but not on control transfectants. Detects Glut2 on the surface of the human intestinal cell line Caco2 in flow cytometry and on fixed cells in immunocytochemistry tests. Based on flow cytometric tests on transfected cells, this antibody has no crossreactivity with hGlut1 or hGlut3.
Glucose transporter 2 isoform is an integral plasma membrane glycoprotein of the liver, islet beta cells, intestine, and kidney epithelium. It mediates facilitated bidirectional glucose transport. Because of its low affinity for glucose, it has been suggested as a glucose sensor.

SLC2A2 antibody can be used in ELISA, Western Blot starting at 1 - 2 μg/mL, immunohistochemistry starting at 5 μg/mL, immunofluorescence, and immunoprecipitation.

Protein G Column

PBS, 5% Trehalose

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SLC2A2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human